Learn More
Biodesign Inc of Ny Sieve Holder, two piece plastic with meteal tightener, set of all 5 sizes
Supplier:  Biodesign Inc of Ny F101
Encompass
Sieve Holder used to tightly hold CellMicroSieve filters to form a funnel through which extraction liquid can easily be collected
Catalog No. NC2684796