Learn More
Cole-Parmer Essentials Sieve; No 18, Full-Height, 8", SS/SS, NIST-Traceable Calibration Certificate
Supplier:  Cole-Parmer 5998455
Encompass_Preferred
Sieve is serialized, comes with a test sieve certificate, and meets ASTM E11 specifications. Includes an NIST-traceable calibration report supplied by the manufacturer.
Catalog No. NC1969916