Learn More
Cosmo Bio Usa Inc SIG IMMUNOREACTION SOL 250ML
SDP
Supplier:  Cosmo Bio Usa Inc TYBNKB101
Encompass
Antibody signal enhancer solution set for 1 (250ml) and 2 (250ml) antibody. Simply dilute Ab to improve sensitivity, background, S/N ratio in most applications. Saves ab. Colorimetric, chemiluminescent, fluorescent compatible.
Catalog No. NC9835260