Learn More
Sigma Aldrich Fine Chemicals Biosciences SIG10 5a Chemically Compet
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CMC000720X40UL
Encompass
The SIG10 5a Chemically Competent Cells share the most useful genetic elements of standard cloning strains like DH5alpha(TM) and directly replace them in cloning protocols. They provide the performance researchers need with ease of use as only common laboratory equipment is required and provide SOLUTIONons for a wide range of applications at economical prices.GenotypefhuA2Delta(argf-lacZ)U169 phoA glnV44 Phi80 Delta(lacZ)M15 gyrA96 recA1 relA1 endA1 thi-1 hsdR17
Catalog No. 50-290-5005