Learn More
Medchemexpress LLC Siglec-E Mouse HEK 500ug
SDP
Supplier:  Medchemexpress LLC HYP71314500UG
Encompass_Preferred
Recombinant mouse Siglec-E extracellular domain fused to a human Fc, expressed in HEK293 cells and supplied as a lyophilized powder for use in binding assays, receptor-ligand studies, and antibody development. Certificate of analysis supports purity and handling recommendations.
  • Expressed in HEK293 cells to preserve mammalian glycosylation.
  • Human Fc fusion enables easy detection and purification.
  • High purity (≥98%) as confirmed by SDS-PAGE.
  • Lyophilized format for long-term storage and transport.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
  • Sequence corresponds to the extracellular region (Q20-F355).
  • Stable when stored at -20°C; aliquot and add carrier for extended storage.
Catalog No. 50-001-96739