Learn More
Sigma Aldrich Fine Chemicals Biosciences SIGMAFAST DAB with Metal Enhancer, 50 set
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D042650SET
Encompass
Tablet, storage temp: −20°C, Synonym: 3,3′-Diaminobenzidine Peroxidase Substrate, DAB Peroxidase Substrate. SIGMAFAST DAB with Metal Enhancer has been used in the visualization of lung tissue sections using immunohistochemistry, dorsal root ganglion (DRG) neurons and immunohistochemical staining for collagen in anterior cruciate ligament (ACL) cells.
Catalog No. NC0046673