Learn More
Agilent Technologies SIGNAL 2-AB-PLUS LAB.KIT
Supplier:  Agilent Technologies GKK804
Encompass
The Signal 2-AB-plusLabeling Kit improves on the traditional 2-AB Labeling Kit(GKK-404)by providing the 2-AB and Reductant already in solution. You can now use what you need, and store the rest of the kit for later use. No more glass ampoules! We also increased the pack size for the Signal 2-AB-plusLabeling Kit, allowing you to process up to 48 samples.
Catalog No. NC0876052