Learn More
Medchemexpress LLC Signal recognition particle 54 (SRP54) protein | 10 UG

Supplier: Medchemexpress LLC HYP7453710UG
SRP54 is a recombinant human signal recognition particle 54 kDa protein produced in Sf9 insect cells and provided with a histidine purification tag. The protein is supplied as a filtered, buffered solution formulated for research applications, with recommended frozen storage to preserve biochemical integrity and low endotoxin to minimize assay interference.
- Recombinant human protein expressed in Sf9 insect cells.
- C-terminal His tag for affinity purification.
- Formulated in 20 mM Tris, pH 7.5, 300 mM NaCl, 10% glycerol; 0.22 μm filtered.
- Approximate molecular weight ~58 kDa (residues M1-M504, accession P61011).
- Low endotoxin level: <1 EU/μg as measured by LAL.
- Store at -80 °C; stable short-term at -20 °C; avoid repeated freeze-thaw.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.