Learn More
ATCC SiHa; Cervical Carcinoma; Human (Homo sapiens)
Supplier:  ATCC HTB35
Encompass
SiHa is a cell line isolated from fragments of a primary uterine tissue sample from a 55-year-old, female, Japanese patient with squamous cell carcinoma. Genes expressed are p53+ and pRB+. This cell line is a suitable transfection host.
Catalog No. 50-238-2880
 Restricted Item