Learn More
SAINT GOBAIN PERF PLASTICS POESTENKIL Solid Silicone Stoppers, U.S. Size No., 2, European Size No., 21D
Supplier:  SAINT GOBAIN PERF PLASTICS POESTENKIL D1069811
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Create a tight, reliable seal in virtually all sealing applications. Pure, high-grade, translucent silicone withstands repeated sterilization, as well as temperatures from -60° to +220°C (-76° to +428°F) peaks at 250°C (482°F). Resist particle buildup Chemically inert and are food-grade qualityIdeal for procedures demanding high purity&mdash ideal for biological applications (after normal cleaning and autoclaving).
Catalog No. 50-206-7761