Learn More
Accuflow Systems Saint-Gobain Versilic Silicone Stopper, 31D, 25.9x32.0x30.0 mm
Supplier:  Accuflow Systems DX26303110
Encompass_Preferred
Versilic silicone stopper designed to provide a secure, chemical-resistant seal for laboratory and bioprocess vessels. Platinum-cured silicone offers low extractables and broad chemical compatibility; folding-skirt geometry ensures reliable sealing on glass and plastic containers. Commonly used in bioprocessing, analytical, and general laboratory closures.
  • Platinum-cured silicone for low extractables and biocompatibility.
  • Folding skirt geometry for a secure seal on glass and plastic vessels.
  • Dimensions: top 25.9 mm, bottom 32.0 mm, height 30.0 mm.
  • Size designation: 31D.
  • Pack quantity: 10 per pack.
Catalog No. NC2327777