Learn More
Waters Corp SILAC Hi3 Phos B Standard, Stable Isotope Labeled, Quantitative Proteomics, Microbial Origin
Supplier:  Waters Corp 186007083
Encompass_Preferred
This stable isotope labeled (SIL) standard is used for quantitative proteomics, specifically designed for performing Hi3 relative protein quantitation in samples of microbial origin. It provides an isotopically unique exogenous standard.
  • Perform relative protein quantitation of mixed proteome samples.
  • Minimize interferences with endogenous peptides.
  • Add a second level of internal quality control (i.e., spike standards toward the lower limit of detection).
  • Provides both a labeled and unlabeled version of the top 6 ionizing peptides represented in each proteome.
  • Intended for samples of microbial origin to minimize interferences between the standard and the protein sample.
  • May also be of use in the evaluation and benchmarking of proteomic LC/MS systems, including time-of-flight mass spectrometers.
Catalog No. 50-239-1466