Learn More
Accuflow Systems Masterflex Precision Pump Tubing, Platinum-Cured Silicone, 0.252 in, 200 ft
Supplier:  Accuflow Systems 9640326
Encompass_Preferred
Precision platinum-cured silicone pump tubing designed for peristaltic pumps. Manufactured to tight dimensional tolerances for accurate, repeatable flow and extended service life. The tubing's smooth inner surface and low-leachable formulation make it suitable for laboratory and process fluid handling where chemical compatibility and purity are important.
  • Platinum-cured silicone construction.
  • 0.252 in (6.40 mm) inner diameter.
  • 200 ft spool length.
  • Smooth inner liner with low leachables.
  • Engineered for consistent flow and long pump life.
  • Suitable for laboratory and process peristaltic pumps.
Catalog No. NC3371714