Learn More
Selleck Chemical LLC Sildenafil S4684-500mg
SDP
Supplier:  Selleck Chemical LLC S4684500MG
Encompass_Preferred
Sildenafil (Revatio UK-92480 Viagra) is a Phosphodiesterase 5 Inhibitor with IC50 of 5 22 nM
Catalog No. 50-311-0120