Learn More
Medchemexpress LLC Sildenafil chlorosul 10g
SDP
Supplier:  Medchemexpress LLC HYW09976110G
Encompass_Preferred
Sildenafil chlorosulfonyl is a synthetic intermediate in the synthesis of the phosphodiesterase 5 (PDE5) inhibitor Sildenafil (HY-15025)
Catalog No. 50-002-06452