Learn More
Medchemexpress LLC Sildenafil (citrate) | 171599-83-0 | C28H38N6O11S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15025A10MM/1ML
Encompass_Preferred
Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It is intended for research use only and is available in solid form or as a 10 mM solution in DMSO. Various quantities are available, and other forms like Sildenafil and deuterated versions also exist.
  • Potent PDE5 inhibitor with an IC50 of 5.22 nM.
  • Suitable for research purposes.
  • Available in various forms and quantities, including solid or 10 mM solution in DMSO.
  • Cited in numerous top scientific publications.
  • Biological activity demonstrated in vitro and in vivo.
  • Associated with numerous clinical trials for various conditions.
Catalog No. 50-003-75314