Learn More
TARGETMOL CHEMICALS INC Sildenafil citrate 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0467500MG
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Sildenafil citrate (UK-92480 citrate) , a cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5) Inhibitor, is used extensively for erectile dysfunction and less commonly for pulmonary hypertension. Purity 99.59%
Catalog No. 50-288-0358