Learn More
Abcam Sildenafil citrate, phosphodiesterase 5 (PDE5) inhibitor, 10MG
Supplier:  Abcam AB14196510MG
Encompass_Preferred
MW 666.7 Da, Purity >99%. Potent, orally active phosphodiesterase 5 (PDE5) inhibitor (IC₅₀ = 4 nM). Various pharmacological effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6393