Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sildenafil-d3 | 1126745-90-1 | C22H27D3N6O4S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY15025S5MG

Encompass_Preferred

Sildenafil-d3 is a deuterium-labeled version of Sildenafil, a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. This compound is suitable for various analytical applications.

  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Appearance: solid.
  • Color: white to off-white.
  • Purity: 99.87%.
  • Target: PDE5.
  • Molecular weight: 477.59.

Catalog No. 50-003-72487


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.