Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc SILICA 3UM 120A 150X4.6MM
SDP

Supplier:  YMC America, Inc SL12S031546WT

Encompass_Preferred

Silica 3Um; 120A; 150 X 4.6mm

Catalog No. 50-874-664


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.