Learn More
Sorbent Technologies Inc Silica Gel, Technical Grade, 60A, 40-63um, 25kg
SDP
Supplier:  Sorbent Technologies Inc 4093025KG
Encompass_Preferred
Silica Gel, Technical Grade is an economical choice for general separations and filtration. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 25kg. This silica gel gives good chromatographic performance for general, non-critical separations. It is used for the packing of traditional flash chromatography columns.
Catalog No. NC2397611
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.