Learn More
Miles Scientific SILICA GEL HLF 5X20 25/PLATES
SDP
Supplier:  Miles Scientific P47531
Encompass_Preferred
Silicagel HL and HLF, Silicagel HLF, TLC platesglass-backed UNIPLATES, 250um, 5 x 20cm scored (2.5 x 5cm), 25/BX
Catalog No. NC9264126