Learn More
Electron Microscopy Sciences Silica Gel Packets 10 Gram, 2¾" x 3", 91.5 cubic inch Capacity
SDP
Supplier:  Electron Microscopy Sciences 7120610
Encompass_Preferred
Silica Gel-Desiccant is a fast and extra moisture absorbing agent. Meets FDA specifications for food and drug product. Provides superior absorption. Remains dry at maximum saturation. Silica gel can absorb 15% of its weight in water vapor in two hours.
Dimensions: 2¾" x 3"
Capacity: 91.5 cubic inch
Catalog No. 50-930-892