Learn More
Electronic Imaging Materials Inc 676 SiliCode™ White Polyester for Siliconized Surfaces, 1.50 inch x 1.00 inch, ROLL of 2500, 3 inch core
SDP
Supplier:  Electronic Imaging Materials Inc 676150100
Encompass_Preferred
676 SiliCode™ 1.50 inch x 1.00 inch, White Polyester Labels for Siliconized Surfaces. Gloss thermal transfer material offers a smooth surface and excellent stability in a variety of temperatures. Superior resistance to moisture, scratches, chemicals and UV light. The key feature of this product is its adhesive that is designed to work on silicone-coated surfaces as well as contaminated surfaces. This adhesive will work on a variety of other surfaces including textured and low-surface-energy substrates. Works best with T84 or T68 premium resin ribbon. Good conformability eliminates edge lifting in applications with small radius curves such as airbags and hoses. Will accept flexographic color printing.
Catalog No. NC1399572