Learn More
Electron Microscopy Sciences EMS Silicon Monoxide Film on on 400 mesh Nickel Grid
SDP
Supplier:  Electron Microscopy Sciences SF400NI
Encompass
A thin film of pure silicone Monoxide (15 – 30 nm) is deposited directly on top of the grid. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Catalog No. 50-331-00