Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Platinum-Cured, 0.50 in ID, 0.88 in OD
Supplier:  Accuflow Systems ULTRAC5005F
Encompass_Preferred
Sani-Tech Ultra-C platinum-cured silicone tubing provides ultra-low extractables and long pump life for biopharmaceutical fluid handling. The tubing is supplied as a 50 ft coil with a 0.50 in inner diameter and a 0.88 in outer diameter, and supports a wide temperature range for versatile process use.
  • Platinum-cured silicone for ultra-low extractables and extended pump life.
  • 0.50 in inner diameter, 0.88 in outer diameter, 0.19 in wall, 50 ft coil.
  • Wide temperature range: -80 to 500 °F (-62 to 260 °C) for harsh process conditions.
  • Tested to USP <88> Class VI, USP <87>, ISO 10993-5, and European Pharmacopoeia 3.1.9 for bioprocess suitability.
  • Suitable for sterile transfer, media and buffer processing, pumping, cell culture, chromatography, and final fill operations.
Catalog No. NC3312437