Learn More
Electron Microscopy Sciences Silicone Carbide Powder, 14 µm particles 1 LB
SDP
Supplier:  Electron Microscopy Sciences 5026014
Encompass_Preferred
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 14 µm
Catalog No. 50-997-176