Learn More
Med Vet International Silicone Nasogastric Feeding Tube, J0386A, 5.0Fr, 1.7mm x 20cm, Each
Supplier: Med Vet International JOR386A
These silicone feeding tubes are great for administering medication and other fluids orally or nasally. All catheters are fully autoclavable. Lateral eyes are on distal tip. Hinged-attached cap.