Learn More
Medchemexpress LLC HY-50855 10mg Medchemexpress, Silmitasertib CAS:1009820-21-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5085510MG
Encompass_Preferred
Medchemexpress, HY-50855 10mg Silmitasertib CAS:1009820-21-6 Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9339