Learn More
Selleck Chemical LLC Silmitasertib (CX-4945) 10mM/1mL 1009820-21-6
SDP
Supplier:  Selleck Chemical LLC S224810MM/1ML
Encompass_Preferred
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay, less potent to Flt3, Pim1 and CDK1 (inactive in cell-based assay). Silmitasertib induces autophagy and promotes apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2208