Learn More
Trajan Scientific SilTite Union Micro Medical 1/EA
SDP
Supplier:  Trajan Scientific 073561
Encompass_Preferred
SilTite -union for joining 0 1 - 0 25 mm and 0 32 mm columns/fused silica
Catalog No. 50-004-94266