Learn More
Sigma Aldrich Fine Chemicals Biosciences SILuTMMAb Nivolumab Stable
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MSQC27100UG
Encompass
SILu(TM)MAb Nivolumab (MSQC27) is a recombinant stable isotope-labeled monoclonal antibody which incorporates [13C6 15N4]-Arginine and [13C6 15N2]-Lysine. Expressed in CHO cells it is designed to be used as an internal standard for the quantitative mass spectrometry analysis of Nivolumab in human and animal serum. SILu(TM)MAb Nivolumab is for research use only and is not intended for diagnostic or therapeutic use.
Catalog No. 50-290-4609