Learn More
Hampton Research Silver Bullets, 0.5ml
SDP
Supplier:  Hampton Research HR2096
Encompass
Silver Bullets screens a portfolio of small molecules for their ability to establish stabilizing, inter molecular, hydrogen bonding, hydrophobic and electrostatic interactions which could promote stability, lattice formation, and crystallization. Please note items from this supplier are non-returnable.
Catalog No. 50-260-3