Learn More
Perkin Elmer US LLC Silver Calibration Reference Standard
Supplier:  Perkin Elmer US LLC N5370438
Encompass_Preferred
PerkinElmer reference standards are 99.999% pure. DSC, STA, and TMA instruments require elements of this purity to ensure these analyzers are accurately calibrated.
Catalog No. 50-211-1451