Learn More
Medchemexpress LLC Silver chloride | 7783-90-6 | MFCD00003399 | 99.5% | 143.32 | AgCl | 25 G

Supplier: Medchemexpress LLC 25GHYW019929
Silver chloride (AgCl) is a high-purity inorganic reagent (99.5%) used in analytical and life-science research. It is employed as a reference material and reagent in spectroscopic and electrochemical applications, providing consistent composition and reliable performance for laboratory workflows.
- Purity 99.5% for analytical consistency.
- Chemical formula AgCl and molecular weight 143.32.
- CAS number 7783-90-6 for unambiguous identification.
- Low solubility in water suitable for precipitation and reference uses.
- Delivered as a stable solid for easy handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.