Learn More
Medchemexpress LLC Silver(I) chloride | 7783-90-6 | MFCD00003399 | 99.5% | 143.32 g/mol | AgCl | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0199295G
Encompass_Preferred
Silver chloride (AgCl) is an inorganic reagent-grade compound at 99.5% purity. This white crystalline solid is commonly used in analytical chemistry, electrochemistry, and life-science research as a reagent, reference material, or for electrode preparation. Intended for research use only.
  • High purity (99.5%) suitable for analytical applications.
  • White crystalline solid with well-defined composition (AgCl).
  • Low solubility useful for precipitation and electrode preparation.
  • Consistent molecular weight supports quantitative analyses.
  • Available in multiple pack sizes to fit laboratory workflows.
Catalog No. 50-002-99998