Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Silver Nitrate 0.0141 Normal NIST Traceable, Solution (1L)
SDP

Supplier:  Aqua Solutions 83701L

Encompass_Preferred

Silver Nitrate 0.0141 Normal NIST Traceable, Solution (1L)

Catalog No. NC0663731


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.