Learn More
Medchemexpress LLC Silver, [4-amino-N-(2-pyrimidinyl-κN1)benzenesulfonamidato-κO]- | 22199-08-2 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB14971G
Encompass_Preferred
Silver sulfadiazine (AgSD) is a sulfonamide antibiotic that exerts a dual inhibitory action on bacterial growth. Its sulfa moiety (SD-SDZ) prevents bacterial folate absorption and subsequent DNA synthesis. Additionally, the silver released from Silver sulfadiazine binds and disrupts the DNA structure, thereby precluding bacterial DNA replication.
  • Sulfonamide antibiotic
  • Exerts dual inhibitory action on bacterial growth
  • Prevents bacterial folate absorption and DNA synthesis
  • Disrupts bacterial DNA structure
  • Precludes bacterial DNA replication
  • Used in the research of bacterial infection in burns
Catalog No. 50-004-11955