Learn More
Medchemexpress LLC Silybin | 802918-57-6 | 482.44 | 50 MG

Supplier: Medchemexpress LLC HYN0779A50MG
Silybin is a flavonolignan isolated from milk thistle (Silybum marianum) seeds. It induces apoptosis and exhibits hepatoprotective, antioxidant, anti-inflammatory, and anti-cancer activity.
- Exhibits hepatoprotective properties
- Acts as an antioxidant
- Possesses anti-inflammatory effects
- Demonstrates anti-cancer activity
- Induces apoptosis
- High purity (99.93%)
- White to off-white solid appearance
- Molecular formula: C25H22O10
- Soluble in various solvents for in vitro and in vivo studies
- Store powder at 4°C, protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.