Learn More
Medchemexpress LLC HY-N7046 1mg Medchemexpress, Silybin B CAS:142797-34-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN70461MG
Encompass_Preferred
Medchemexpress, HY-N7046 1mg Silybin B CAS:142797-34-0 Silybin B, a flavonolignan separated from Silybum marianum, has anti-tumor activity. Silybin B is the most potent antifibrillogenic and anti-oligomeric component of silymarin and proposes it as a promising anti Alzheimer’s disease drug candidate [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3980