Learn More
Apexbio Technology LLC Silymarin 22888-70-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N17115.1
Encompass_Preferred
Silymarin (CAS 22888-70-6) is a naturally occurring flavonoid complex derived from the seeds of plants belonging to the Asteraceae family notably milk thistle (Silybum marianum) This phytochemical mixture exerts cellular antioxidant and anti-inflammatory activities primarily by inhibiting lipid peroxidation scavenging free radicals stabilizing cell membranes and modulating intracellular signaling pathways linked to oxidative stress response In biomedical research silymarin is extensively studied for hepatoprotective properties and its potential applications in investigating liver injury hepatotoxicity models and oxidative stress-mediated cellular damage The compound can be appropriately dissolved in solvents such as methanol ethanol acetone or ethyl acetate and requires storage under dry cool and sealed conditions
Catalog No. 50-000-63284