Learn More
Bio X Cell Biosimilar Antibody, Human PD-L1, IgG1, 150 kDa
Supplier: Bio X Cell SIM0009R001MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Atezolizumab, making it suitable for research applications. It targets human PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274, and is designed to block its interaction with PD-1 and CD80, which is important in tumor immune evasion.
- Non-therapeutic biosimilar for research.
- Reacts with human PD-L1, B7-H1, or CD274.
- Molecular weight: 150 kDa.
- Purity: ≥95%.
- Endotoxin: ≤0.5 EU/mg.
- Sterility: 0.2 μm filtration.
- Produced in an animal-free facility.
- Purified by Protein A.
- Formulation: PBS, pH 7.0, no stabilizers or preservatives.
- Applications: in vivo PD-L1 blockade, flow cytometry, western blot.
- Storage: 4°C, do not freeze.
- Aggregation: <5%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.