Learn More
Bio X Cell InVivoSIM anti-human HER2 (Trastuzumab Biosimilar), 25 mg
Supplier:  Bio X Cell SIM0005R025MG
Encompass_Preferred
This biosimilar antibody is designed for preclinical research, offering high purity and low endotoxin levels suitable for in vivo applications. It targets human HER2 and provides a cost-effective solution for researchers studying Trastuzumab.
  • High purity and low endotoxin
  • Preservative-free formulation
  • Suitable for in vivo preclinical research
  • Cost-effective solution for research
  • Targets human HER2
Catalog No. NC1959875