Learn More
Medchemexpress LLC Simethicone | 8050-81-5 | 96.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1095195G
Encompass_Preferred
Simethicone is an orally active defoamer that reduces the surface tension of air bubbles in the gastrointestinal tract, leading to their expulsion by vomiting, exhalation, or absorption into the bloodstream. It has potential applications in flatulence and colic.
  • Reduces surface tension of air bubbles in the gastrointestinal tract.
  • Facilitates expulsion of air bubbles by vomiting, exhalation, or absorption.
  • Potential applications in flatulence and colic.
  • Demonstrated gastrointestinal regulation function in rats.
  • Improved colon permeability and hypersensitivity in rats.
  • Improved image quality scores for the jejunum in rabbits.
Catalog No. 50-003-45084