Learn More
Biomatik Corporation Simian Growth Differentiation Factor 15 (GDF15) ELISA Kit
Supplier:  Biomatik Corporation EKU12355
Encompass_Preferred
Simian Growth Differentiation Factor 15 (GDF15) ELISA Kit for detecting GDF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 0.625-40ng/ml. Sensitivity: 0.242ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G7PWZ3. Target Synonyms: PDF; MIC1; NAG-1; PLAB; PTGFB; Macrophage inhibitory cytokine 1; NSAID-activated gene 1 protein; Placental bone morphogenetic protein; Prostate differentiation factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6074