Learn More
Medchemexpress LLC Simmiparib 25mg | 1551355-46-4 | 486.42 g/mol | C23H18F4N6O2 | 25 MG

Supplier: Medchemexpress LLC HY11555225MG
Simmiparib (CAS 1551355-46-4) is a potent, orally active inhibitor of poly(ADP-ribose) polymerase 1 and 2 (PARP1/2) used in preclinical biochemical and cellular research.
- High purity: 99.01%.
- Molecular formula C23H18F4N6O2 and molecular weight 486.42 g/mol.
- Supplied as a 25 mg solid and as 10 mM 1 mL DMSO stock solutions.
- Soluble in DMSO, suitable for preparing assay stocks.
- Intended for PARP1/2 inhibition and DNA damage response studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.