Learn More
AdipoGen Simocyclinone D8
SDP
Supplier:  AdipoGen BVT0290M001
Encompass_Preferred
Chemical. CAS 301845-97-6. Formula C46H42ClNO18. MW 932.3. Isolated from Streptomyces antibioticus Tü 6040. Antibiotic. Bacterial DNA gyrase inhibitor Staphylococcus aureus IC50= 1.45µM, E. coli IC50= 0.41µM. Human topoisomerase II Topo II inhibitor IC50= 100µM. Antibacterial Gram-positive. Antitumor compound.
Catalog No. 50-256-2010