Learn More
Medchemexpress LLC Simotinib | 944258-89-3 | 99.4% | 500.95 | C25H26ClFN4O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10182050MG
Encompass_Preferred
Simotinib is a selective, specific, orally bioavailable EGFR tyrosine kinase inhibitor intended for research use. It inhibits EGFR with an IC50 of 19.9 nM and exhibits antineoplastic activity. Provided as a high-purity analytical standard for biochemical and cellular studies.
  • Selective EGFR tyrosine kinase inhibition (IC50 19.9 nM).
  • Orally bioavailable compound suitable for in vitro and preclinical research.
  • High purity analytical standard (~99.4%).
  • Molecular weight 500.95 and formula C25H26ClFN4O4 for accurate identification.
  • Available as a 50 MG research quantity.
Catalog No. 50-002-48497