Learn More
Medchemexpress LLC Simotinib | 944258-89-3 | 99.4% | 500.95 g/mol | C25H26ClFN4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10182010MG
Encompass_Preferred
Simotinib is a selective, orally bioavailable epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor with reported antineoplastic activity and an IC50 of 19.9 nM. The compound has molecular weight 500.95, reported purity 99.39%, and CAS number 944258-89-3.
  • Selective EGFR inhibition with IC50 19.9 nM.
  • Orally bioavailable kinase inhibitor.
  • High reported purity (99.39%).
  • Molecular weight 500.95 for formulation calculations.
  • Reported antineoplastic activity in preclinical studies.
Catalog No. 50-002-48596