Learn More
Medchemexpress LLC Simotinib | 944258-89-3 | MFCD34469138 | 99.4% | 500.95 g/mol | C25H26ClFN4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10182025MG
Encompass_Preferred
Simotinib is a selective, orally bioavailable epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor provided as an analytical standard for research use. It exhibits antineoplastic activity and is characterized by high purity and defined chemical properties, making it suitable for assay development, analytical reference, and preclinical studies.
  • High potency against EGFR with reported IC50 of 19.9 nM.
  • High purity suitable for analytical and research applications.
  • Supplied in a 25 mg quantity convenient for small-scale studies.
  • Chemical formula C25H26ClFN4O4; molecular weight 500.95 g/mol.
  • Intended for use as a reference standard in assay development and compound characterization.
Catalog No. 50-002-48563